Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ 4 Point Chin Strap for SecureFit™ Safety Helmet

Catalog No. 19071201 Shop All 3M Products
Change view
Click to view available options
Includes:
Buckle
Magnetic buckle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-071-201 Buckle
19-071-200 Magnetic buckle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-071-201 Supplier 3M™ Supplier No. 7100207831

missing translation for 'validMainframeMsg'

Designed with quick and smooth adjustment features for a customized fit.

  • Easy adjustment
  • Fits 3M™ SecureFit™ X5000 Series Safety Helmets
  • 3M Catalog# X5-P4PTCS1 , 3M Catalog# X5-S4PTCS1

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) 3M SecureFit X5000 Series Safety Helmets
For Use With (Application) Construction, Heavy Infrastructure, Oil & Gas
Includes Buckle
Material Nylon
Model X5-S4PTCS1
Product Type Chin Strap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.