Learn More
Chemglass Life Sciences 4-Port Adapter, GL-80, (2) 1/4" & (2) 1/2" Hose Barbs, Stainless Steel
SDP
Supplier:  Chemglass Life Sciences CLS1380480
Encompass
The 4-Port inlet assembly fits bottles with GL-80 threads.The assembly incorporates four separate pipes into one assembly for efficient use of space.Each port can be used individually for reagent addition.Adapter has (2) 1/4 Hose and (2) barbs for connection to flexible tubing.Inside the reactor, the tubes are 1 long and are barbed.
Catalog No. 50-273-3321