Learn More
Chemglass Life Sciences 4-Port Adapter, M27 Thread, (4) 1/4" Angled Hose Barbs, Stainless Steel
SDP
Catalog No. 502733322
Encompass
The 4-Port inlet assembly fits into M27 head plate port.The assembly incorporates four separate pipes into one assembly for efficient use of space.Each port can be used individually for reagent addition.M27 threaded adapter has (4) 1/4 hose barbs for connection to flexible tubing.Inside the reactor, the 1/4 tubes are (2) @.75, (1)@.96 and (1)@1.16 long.
Catalog No. 50-273-3322 Supplier Chemglass Life Sciences Supplier No. CLS1380404
The 4-Port inlet assembly fits into M27 head plate port.The assembly incorporates four separate pipes into one assembly for efficient use of space.Each port can be used individually for reagent addition.M27 threaded adapter has (4) 1/4 hose barbs for connection to flexible tubing.Inside the reactor, the 1/4 tubes are (2) @.75, (1)@.96 and (1)@1.16 long.