Learn More
Chemglass Life Sciences Vacuum Manifold, Right Handed, 4 Ports, 0-8mm Chem-Cap Valve, 345mm Overall width
SDP
Supplier:  Chemglass Life Sciences CG443404
Encompass_Preferred
Vacuum Manifold, Right Handed, 4 Ports, 0-8mm Chem-Cap Valve, 345mm Overall width. General purpose single bank vacuum manifold with 4 ports. Main tube is 38mm OD with a serrated hose connection, having the smallest serration 13.5mm and the largest serration 15.5mm, located at either the left or right side for vacuum connection. Ports are 0-8mm Chem-Vac™ Chem-Cap valves with serrated hose connections. The 0-8mm has the smallest serration 13.5mm and the largest serration 15.5mm. Ports are spaced 75mm center to center for the 0-8mm size.
Catalog No. CG443404