Learn More
Ace Glass, Inc. 4 PORT UHDPE HDL MANIFOLD 4 PORT UHDPE HDL MANIFOLD
SDP
Catalog No. 50979871
Encompass
4 port vacuum/ glass manifold with a Firestone Rapid Purge Valve at one end and 0-5mm high vacuum easy action PTFE stopcock ports with 3/8¿ O.D. Swagelok tubing connections. Valve eliminates the need for a second tube, and includes shut-off float on the gas side to prevent pressure buildup when system is purged. Each connection has an o-ring groove for use with #11 Ace-Thred. Plugs are PTFE with UHDPE handles and FETFE o-rings.
Catalog No. 50-979-871 Supplier Ace Glass, Inc. Supplier No. 876527
4 port vacuum/ glass manifold with a Firestone Rapid Purge Valve at one end and 0-5mm high vacuum easy action PTFE stopcock ports with 3/8¿ O.D. Swagelok tubing connections. Valve eliminates the need for a second tube, and includes shut-off float on the gas side to prevent pressure buildup when system is purged. Each connection has an o-ring groove for use with #11 Ace-Thred. Plugs are PTFE with UHDPE handles and FETFE o-rings.