Learn More
Chemglass Life Sciences 4-Port Vacuum Manifold, Double Bank, Airfree/Inert Line, Schlenk, Complete
SDP
Supplier:  Chemglass Life Sciences AF045201
Encompass_Preferred
4-Port Vacuum Manifold, Double Bank, Airfree/Inert Line, Schlenk, Complete. Flexible and compact, this 4-port line is constructed using CG-474-02, 4mm oblique hollow high vacuum greased stopcocks on all four ports. Ports are fitted with two 14/20 inner joints and two 24/40 inner joints spaced 5" center to center. This configuration permits use directly with vessels having these joint sizes or the AF-0455 adapters to fit standard flexible hose. All four ends have #15 o-ring joints permitting the trap assembly to be attached to the right or left side of the vacuum bank. Manifold is supplied with two AF-021-001 #15 stoppers and three AF-007-005 #15 o-ring joint to serrated hose adapters. Additional adapters can be found in the catalog under AF-0070 through AF-0080, which enables the manifold to be set up using any number of variations.
Catalog No. AF045201