Learn More
LUNA NANOTECH INC 4-Positions Maxi Magnetic Rack For 50 ML Tubes
Catalog No. 502566964
Encompass_Preferred
Convenient MAXI format magnetic racks for large scale separation of magnetic beads in 50 mL centrifuge tubes. These racks can be used with our PuroMAG MAXI kits as well as various magnetic beads offered by Luna Nanotech and other manufactureres.
Catalog No. 50-256-6964 Supplier LUNA NANOTECH INC Supplier No. MGRM004
Convenient MAXI format magnetic racks for large scale separation of magnetic beads in 50 mL centrifuge tubes. These racks can be used with our PuroMAG MAXI kits as well as various magnetic beads offered by Luna Nanotech and other manufactureres.