Learn More
Medchemexpress LLC 4-PPBP maleate | 201216-39-9 | 99.6% | 409.52 | C25H31NO4 | 10 MG
SDP
Catalog No. 5000212583
Encompass_Preferred
4-PPBP maleate is a research-grade sigma-1 (σ1) receptor ligand and agonist that also acts as a non-competitive, selective NR1a/2B NMDA receptor antagonist. It is provided as the maleate salt in small-mass vials for in vitro and neuroprotection research; handle according to the safety data sheet.
Catalog No. 50-002-12583 Supplier Medchemexpress LLC Supplier No. HY10104310MG
4-PPBP maleate is a research-grade sigma-1 (σ1) receptor ligand and agonist that also acts as a non-competitive, selective NR1a/2B NMDA receptor antagonist. It is provided as the maleate salt in small-mass vials for in vitro and neuroprotection research; handle according to the safety data sheet.