Learn More
Medchemexpress LLC 4-PPBP maleate | 201216-39-9 | 99.6% | 409.52 g/mol | C25H31NO4 | 50 MG
SDP
Catalog No. 5000213234
Encompass_Preferred
4-PPBP maleate is a research-grade sigma-1 (σ1) receptor ligand and agonist used in neuroscience studies. It acts as a non-competitive, selective antagonist of NR1a/2B NMDA receptors and has been used in models of neuroprotection. The compound is supplied as a solid with high reported purity and defined storage recommendations for stability.
  • Potent sigma-1 receptor ligand and agonist.
  • Non-competitive, selective NR1a/2B NMDA receptor antagonist.
  • Used in neuroprotection research models.
  • High purity (99.6%).
  • Supplied as a solid for convenient handling.
  • Stable when stored sealed at 4°C; solvent storage conditions applicable.
Catalog No. 50-002-13234 Supplier Medchemexpress LLC Supplier No. HY10104350MG
4-PPBP maleate is a research-grade sigma-1 (σ1) receptor ligand and agonist used in neuroscience studies. It acts as a non-competitive, selective antagonist of NR1a/2B NMDA receptors and has been used in models of neuroprotection. The compound is supplied as a solid with high reported purity and defined storage recommendations for stability.
  • Potent sigma-1 receptor ligand and agonist.
  • Non-competitive, selective NR1a/2B NMDA receptor antagonist.
  • Used in neuroprotection research models.
  • High purity (99.6%).
  • Supplied as a solid for convenient handling.
  • Stable when stored sealed at 4°C; solvent storage conditions applicable.