Learn More
Medchemexpress LLC 4-PPBP maleate | 201216-39-9 | MFCD00209909 | 99.6% | 409.52 g·mol-1 | C25H31NO4 | 25 MG
SDP
Catalog No. 5000212825
Encompass_Preferred
4-PPBP maleate is a research-grade small-molecule sigma-1 receptor ligand and NMDA receptor modulator supplied as the maleate salt. It is intended for in vitro and preclinical studies and is not for human or clinical use. The compound is well characterized and provided with analytical purity suitable for laboratory research.
  • Potent sigma-1 receptor ligand and agonist.
  • Selective, non-competitive modulator of NR1a/2B NMDA receptors.
  • Supplied as the maleate salt for stability and handling.
  • High purity suitable for research applications (approximately 99.6%).
  • Well-characterized molecular identity: C25H31NO4; MW 409.52 g·mol-1.
  • Available in small laboratory pack sizes (25 mg).
Catalog No. 50-002-12825 Supplier Medchemexpress LLC Supplier No. HY10104325MG
4-PPBP maleate is a research-grade small-molecule sigma-1 receptor ligand and NMDA receptor modulator supplied as the maleate salt. It is intended for in vitro and preclinical studies and is not for human or clinical use. The compound is well characterized and provided with analytical purity suitable for laboratory research.
  • Potent sigma-1 receptor ligand and agonist.
  • Selective, non-competitive modulator of NR1a/2B NMDA receptors.
  • Supplied as the maleate salt for stability and handling.
  • High purity suitable for research applications (approximately 99.6%).
  • Well-characterized molecular identity: C25H31NO4; MW 409.52 g·mol-1.
  • Available in small laboratory pack sizes (25 mg).