Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. P020625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P020625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P020625G Supplier TCI America Supplier No. P020625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5438-19-7
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00013989
InChI Key GDFUWFOCYZZGQU-UHFFFAOYSA-N
Synonym p-propoxybenzoic acid, 4-n-propoxybenzoic acid, benzoic acid, 4-propoxy, 4-n-propyloxybenzoic acid, 4-propoxy-benzoic acid, benzoic acid, p-propoxy, p-propyloxybenzoic acid, pubchem2681, 4-propoxy benzoic acid, p-carboxyphenoxy propane
PubChem CID 138500
IUPAC Name 4-propoxybenzoic acid
SMILES CCCOC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 144°C
Color White
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Propoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.