Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propoxybenzonitrile, 97%, Thermo Scientific™

Catalog No. BTB09860DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB09860DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB09860DA Supplier Thermo Scientific Chemicals Supplier No. BTB09860DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60758-84-1
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00173874
InChI Key VYOPNUZYNSNYRA-UHFFFAOYSA-N
Synonym 4-propoxy-benzonitrile, 4-propoxybenzenecarbonitrile, 4-n-propoxybenzonitrile, benzonitrile, 4-propoxy
PubChem CID 322939
IUPAC Name 4-propoxybenzonitrile
SMILES CCCOC1=CC=C(C=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)2OC6H4CN
Quantity 1 g
Formula Weight 161.2
Percent Purity 97%
Chemical Name or Material 4-Propoxybenzonitrile
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.