Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propoxyphenol 98.0+%, TCI America™
SDP

Catalog No. P095525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P095525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P095525G Supplier TCI America Supplier No. P095525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18979-50-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002335
InChI Key KIIIPQXXLVCCQP-UHFFFAOYSA-N
Synonym phenol, 4-propoxy, 4-n-propoxyphenol, p-propoxyphenol, phenol, p-propoxy, hydroquinone monopropyl ether, p-propoxy phenol, 4-propyloxyphenol, 4-propoxy-phenol, pubchem13204, para-propoxyphenol
PubChem CID 29352
IUPAC Name 4-propoxyphenol
SMILES CCCOC1=CC=C(C=C1)O

Specifications

Melting Point 56°C
Quantity 25 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Propoxyphenol
EINECSNumber (3)-3539
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.