Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propoxyphthalonitrile 95.0+%, TCI America™
SDP

Catalog No. P10705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10705G 5 g
P10701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10705G Supplier TCI America Supplier No. P10705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 106144-18-7
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD00059102
InChI Key RDLZRCSOLOTEPK-UHFFFAOYSA-N
Synonym 1,2-Dicyano-4-propoxybenzene
PubChem CID 588896
IUPAC Name 4-propoxybenzene-1,2-dicarbonitrile
SMILES CCCOC1=CC(=C(C=C1)C#N)C#N

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Propoxyphthalonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.