Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Propyl-4-biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P23875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2387-5G 5 g
P23871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2387-5G Supplier TCI America Supplier No. P23875G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 4 '-Propyl-4-biphenylboronic Acid (contains varying amounts of Anhydride)
CAS 153035-56-4
Color White-Yellow
Molecular Formula C15H17BO2
MDL Number MFCD01114670
Quantity 5 g
InChI Key NOQFUISBLHCDSR-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C2=CC=C(C=C2)CCC)(O)O
IUPAC Name [4-(4-propylphenyl)phenyl]boronic acid
Molecular Weight (g/mol) 240.109
PubChem CID 19042596
Formula Weight 240.11
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.