Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propylbiphenyl, TCI America™
SDP

Catalog No. P121610ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P121610ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P121610ML Supplier TCI America Supplier No. P121610ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10289-45-9
Molecular Formula C15H16
Molecular Weight (g/mol) 196.293
MDL Number MFCD00102114
InChI Key NAYIXKXYHOLMRC-UHFFFAOYSA-N
Synonym 4-propylbiphenyl, 4-propyl-1,1'-biphenyl, 4-n-propylbiphenyl, 4-propyl biphenyl, 4-propyl-1,1'-diphenyl, 1,1'-biphenyl, 4-propyl, 1-phenyl-4-propyl-benzene, 4-n-propyl biphenyl, pubchem2514, ksc492q3t
PubChem CID 2734126
IUPAC Name 1-phenyl-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Color Colorless
Boiling Point 296°C
Quantity 10 mL
Formula Weight 196.29
Physical Form Liquid
Chemical Name or Material 4-Propylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.