Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propyltoluene 99.0+%, TCI America™
SDP

Catalog No. P07495ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07495ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P07495ML Supplier TCI America Supplier No. P07495ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1074-55-1
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00048698
InChI Key JXFVMNFKABWTHD-UHFFFAOYSA-N
Synonym 1-Methyl-4-propylbenzene
PubChem CID 14095
IUPAC Name 1-methyl-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)C

Specifications

Color Yellow
Boiling Point 183°C
Quantity 5 mL
UN Number 3295
Formula Weight 134.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Propyltoluene
EINECSNumber (3)-0015
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.