Learn More
MSC 4 PT HARD HAT SUSPENSION
Catalog No. 5000781089
Encompass
This replacement suspension is designed for hard hats, offering a secure and adjustable fit for worker safety and comfort.
  • Fits standard hat sizes
  • Features a ratchet adjustment type
  • Equipped with a 4-point suspension system
  • Color: black
  • Compatible with Topgard Caps or Hats
  • Overall length of 214 mm (8.425 in)
  • Weight of 0.092 kg (0.203 lb)
Catalog No. 50-007-81089 Supplier MSC Supplier No. 57588568
This replacement suspension is designed for hard hats, offering a secure and adjustable fit for worker safety and comfort.
  • Fits standard hat sizes
  • Features a ratchet adjustment type
  • Equipped with a 4-point suspension system
  • Color: black
  • Compatible with Topgard Caps or Hats
  • Overall length of 214 mm (8.425 in)
  • Weight of 0.092 kg (0.203 lb)