Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Pyrid-3-ylphenyl)methanol, ≥97%, Thermo Scientific™

Catalog No. CC40009DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC40009DE 5 g
CC40009CB 250 mg
CC40009DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC40009DE Supplier Thermo Scientific Chemicals Supplier No. CC40009DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 217189-04-3
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD04116293
InChI Key ZHIJCVCCKVZBHE-UHFFFAOYSA-N
Synonym 4-pyrid-3-ylphenyl methanol, 4-pyridin-3-yl phenyl methanol, 4-pyridin-3-ylphenyl methanol, benzenemethanol,4-3-pyridinyl, 4-pyridin-3-yl-phenyl-methanol, 4-3-pyridyl-phenyl-methanol, 4-3-pyridyl phenyl methan-1-ol, 3-4-hydroxymethyl phenyl pyridine, 3-4-hydroxymethylphenyl pyridine
PubChem CID 1515243
IUPAC Name (4-pyridin-3-ylphenyl)methanol
SMILES C1=CC(=CN=C1)C2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 185.22
Percent Purity 97%
Chemical Name or Material (4-Pyrid-3-ylphenyl)methanol
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.