Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyridazinecarboxylic acid, 95%

Catalog No. p-4813149
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399630010 1 g
AC399630050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399630010 Supplier Thermo Scientific Chemicals Supplier No. 399630010
Only null left
Add to Cart

CAS: 50681-25-9 | C5H4N2O2 | 124.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50681-25-9
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.1
InChI Key JUSIWJONLKBPDU-UHFFFAOYSA-N
Synonym 4-pyridazinecarboxylic acid, 4-carboxypyridazine, 4-pyridazinecarboxylicacid, pubchem9501, acmc-1alr8, pyridazin-4-carboxylic acid, 4-pyridazine-carboxylic acid, ksc269i2f, pyridazin-4-yl formate, 1,2-diazine-4-carboxylic acid
PubChem CID 2761046
IUPAC Name pyridazine-4-carboxylic acid
SMILES C1=CN=NC=C1C(=O)O

Specifications

Melting Point 244.2°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 124.1
Percent Purity 95%
Chemical Name or Material 4-Pyridazinecarboxylic acid
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.