Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Pyridin-2-yl-benzaldehyde | 127406-56-8 | 183.21 | 10 G
SDP

Catalog No. 5000447447
Encompass_Preferred

4-Pyridin-2-yl-benzaldehyde is a biochemical reagent suitable for life science research. It serves as a biological material or organic compound and is recognized as a reactive metabolite of Atazanavir.

  • Used as a biochemical reagent
  • Suitable as a biological material or organic compound for life science research
  • Functions as a reactive metabolite of Atazanavir
  • Appears as a solid
  • Color ranges from off-white to light yellow
  • Soluble in DMSO at 100 mg/mL (545.82 mM), requiring ultrasonication
  • Store at 4°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-004-47447 Supplier Medchemexpress LLC Supplier No. HYW00285510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Pyridin-2-yl-benzaldehyde is a biochemical reagent suitable for life science research. It serves as a biological material or organic compound and is recognized as a reactive metabolite of Atazanavir.

  • Used as a biochemical reagent
  • Suitable as a biological material or organic compound for life science research
  • Functions as a reactive metabolite of Atazanavir
  • Appears as a solid
  • Color ranges from off-white to light yellow
  • Soluble in DMSO at 100 mg/mL (545.82 mM), requiring ultrasonication
  • Store at 4°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.