Learn More
Medchemexpress LLC 4-Pyridin-2-yl-benzaldehyde | 127406-56-8 | 183.21 | 25 G
SDP
Catalog No. 5000448285
Encompass_Preferred
4-Pyridin-2-yl-benzaldehyde is a biochemical reagent used as a biological material or organic compound for life science related research. It also serves as a reactive metabolite of Atazanavir.
  • Suitable for life science research
  • Can be used as a biological material or organic compound
  • Functions as a reactive metabolite of Atazanavir
  • Store at 4°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
Catalog No. 50-004-48285 Supplier Medchemexpress LLC Supplier No. 25GHYW002855
4-Pyridin-2-yl-benzaldehyde is a biochemical reagent used as a biological material or organic compound for life science related research. It also serves as a reactive metabolite of Atazanavir.
  • Suitable for life science research
  • Can be used as a biological material or organic compound
  • Functions as a reactive metabolite of Atazanavir
  • Store at 4°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen