Learn More
Accela Chembio Inc 4-pyridineboronic Acid Pinacol Ester | 25g | 181219-01-2 | MFCD01319051 | 97+% | Shelf Life: 900 Days | Light Sensitive/n2/+4
Catalog No. 50444679
Encompass_Preferred
4-pyridineboronic Acid Pinacol Ester | 25g | 181219-01-2 | MFCD01319051 | 97+% | Shelf Life: 900 Days | Light Sensitive/n2/+4
Catalog No. 50-444-679 Supplier Accela Chembio Inc Supplier No. SY00796925G
4-pyridineboronic Acid Pinacol Ester | 25g | 181219-01-2 | MFCD01319051 | 97+% | Shelf Life: 900 Days | Light Sensitive/n2/+4