Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Pyridinecarboxaldehyde

Catalog No. P62402100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P62402100G 100 g
P6240225G 25 g
P62402500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P62402100G Supplier Sigma Aldrich Supplier No. P62402100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-pyridinecarboxaldehyde can be used for the synthesis of:
  • -a,α-unsaturated amides by coupling with n,n-disubstituted formamides
  • Meso-substituted a3-corroles
  • N-(4-pyridylmethyl)-L-valine as a ligand to construct zinc metal-organic frameworks (zn-mofs)
  • 4α-pyridyl terpyridines, with potential application as anticancer and antimicrobial agents
  • 4-pyridinecarboxaldehyde thiosemicarbazone, as a corrosion inhibitor for mild steel

Specifications

CAS 872-85-5
Density 1.137 g/mL (at 20°C (literature))
Boiling Point 71°C to 73°C (10 mmHg)
Molecular Formula C6H5NO
Refractive Index n20/D 1.544 (literature)
Linear Formula C6H5NO
MDL Number MFCD00006425
Quantity 100 g
Synonym Isonicotinaldehyde
Molecular Weight (g/mol) 107.11
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.