Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyridinecarboxaldehyde 96.0+%, TCI America™
SDP

Catalog No. I0143500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0143500ML 500 mL
I014325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0143500ML Supplier TCI America Supplier No. I0143500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 872-85-5
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.112
MDL Number MFCD00006425
InChI Key BGUWFUQJCDRPTL-UHFFFAOYSA-N
Synonym 4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde
PubChem CID 13389
IUPAC Name pyridine-4-carbaldehyde
SMILES C1=CN=CC=C1C=O

Specifications

Boiling Point 187°C
Quantity 500 mL
Formula Weight 107.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Pyridinecarboxaldehyde
RTECSNumber NR9400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.