Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-pyridinecarboxylic acid, 2-[[[2-[[2-(dimethylamino)ethyl]...]]-, hydrochloride | 3026728-26-4 | 96.6% | 25 MG
SDP

Catalog No. 5000231786
Encompass_Preferred

KDM5-C49 hydrochloride is a potent, selective inhibitor of KDM5 histone demethylases used for biochemical and cellular research, including cancer studies. It is supplied as a high-purity solid-liquid mixture with defined storage recommendations to maintain stability.

  • Potent KDM5 inhibitor with reported IC50s of 40 nM (KDM5A), 160 nM (KDM5B), and 100 nM (KDM5C).
  • High purity suitable for research applications.
  • Molecular weight approximately 344.84 g/mol.
  • Physical form: solid-liquid mixture, colorless to light yellow.
  • Recommended storage: 4°C sealed; stock solutions: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small milligram pack sizes for laboratory use.

Catalog No. 50-002-31786 Supplier Medchemexpress LLC Supplier No. HY119397A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KDM5-C49 hydrochloride is a potent, selective inhibitor of KDM5 histone demethylases used for biochemical and cellular research, including cancer studies. It is supplied as a high-purity solid-liquid mixture with defined storage recommendations to maintain stability.

  • Potent KDM5 inhibitor with reported IC50s of 40 nM (KDM5A), 160 nM (KDM5B), and 100 nM (KDM5C).
  • High purity suitable for research applications.
  • Molecular weight approximately 344.84 g/mol.
  • Physical form: solid-liquid mixture, colorless to light yellow.
  • Recommended storage: 4°C sealed; stock solutions: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small milligram pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.