Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-pyridinecarboxylic acid dimethylaminoethyl ethylamino derivative, hydrochloride | 3026728-26-4 | 98.2% | 50 MG
SDP

Catalog No. 5000233154
Encompass_Preferred

KDM5-C49 hydrochloride is a small-molecule inhibitor of KDM5 family histone demethylases used for biochemical and cell-based research to probe demethylase activity.

  • Potent KDM5 demethylase inhibitor.
  • Purity 98.2% (HPLC, batch COA).
  • Molecular weight 344.84 Da.
  • Chemical formula C15H25ClN4O3.
  • Appearance: solid-liquid mixture.
  • Recommended storage: 4°C (sealed, away from moisture); in solvent -80°C (6 months), -20°C (1 month).
  • Typical pack size: 50 MG.

Catalog No. 50-002-33154 Supplier Medchemexpress LLC Supplier No. HY119397A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KDM5-C49 hydrochloride is a small-molecule inhibitor of KDM5 family histone demethylases used for biochemical and cell-based research to probe demethylase activity.

  • Potent KDM5 demethylase inhibitor.
  • Purity 98.2% (HPLC, batch COA).
  • Molecular weight 344.84 Da.
  • Chemical formula C15H25ClN4O3.
  • Appearance: solid-liquid mixture.
  • Recommended storage: 4°C (sealed, away from moisture); in solvent -80°C (6 months), -20°C (1 month).
  • Typical pack size: 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.