Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-pyridinepropanoic acid, α-amino-, dihydrochloride (αR) | 174096-41-4 | MFCD09842075 | 99.8% | 239.10 | C8H12Cl2N2O2 | 10 G
SDP

Catalog No. 5000301580
Encompass_Preferred

H-D-4-Pal-OH.2HCl is the dihydrochloride salt of D-4-pyridylalanine, an alanine derivative provided as a research reagent. It is supplied as a solid with high reported purity and is accompanied by analytical documentation for research use. Store as recommended to maintain stability.

  • High reported purity (99.8%).
  • Solid form, off-white to light yellow.
  • Molecular weight 239.10 g/mol and formula C8H12Cl2N2O2.
  • Dihydrochloride salt suitable for research applications.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent store frozen.
  • Analytical documentation available: datasheet, COA, SDS.

Catalog No. 50-003-01580 Supplier Medchemexpress LLC Supplier No. HYW02240410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-D-4-Pal-OH.2HCl is the dihydrochloride salt of D-4-pyridylalanine, an alanine derivative provided as a research reagent. It is supplied as a solid with high reported purity and is accompanied by analytical documentation for research use. Store as recommended to maintain stability.

  • High reported purity (99.8%).
  • Solid form, off-white to light yellow.
  • Molecular weight 239.10 g/mol and formula C8H12Cl2N2O2.
  • Dihydrochloride salt suitable for research applications.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent store frozen.
  • Analytical documentation available: datasheet, COA, SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.