Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyridinepropanol 97.0+%, TCI America™
SDP

Catalog No. P03515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03515G 5 g
P035125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P03515G Supplier TCI America Supplier No. P03515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2629-72-3
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00047457
InChI Key PZVZGDBCMQBRMA-UHFFFAOYSA-N
Synonym 4-(3-Hydroxypropyl)pyridine, 3-(4-Pyridyl)propanol
PubChem CID 72923
IUPAC Name 3-pyridin-4-ylpropan-1-ol
SMILES C1=CN=CC=C1CCCO

Specifications

Boiling Point 289°C
Quantity 5 g
Formula Weight 137.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Pyridinepropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.