Learn More
Medchemexpress LLC 4-Pyridoxic acid-d3 | 1435934-03-4 | 98.8% | 186.18 | 1 MG
SDP
Catalog No. 5000436713
Encompass_Preferred
4-Pyridoxic acid-d3 is the deuterium labeled 4-Pyridoxic acid, which is a catabolic product of vitamin B6 and is excreted in the urine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-36713 Supplier Medchemexpress LLC Supplier No. HY113493S11MG
4-Pyridoxic acid-d3 is the deuterium labeled 4-Pyridoxic acid, which is a catabolic product of vitamin B6 and is excreted in the urine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.