Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-pyridoxic acid-d3 | 1435934-03-4 | 98.8% | 186.18 g/mol | C8H6D3NO4 | 5 MG
SDP

Catalog No. 5000263571
Encompass_Preferred

4-Pyridoxic acid-d3 is the deuterium-labeled analog of 4-pyridoxic acid, a catabolic product of vitamin B6. It is provided as a stable isotope reference standard for use in analytical, pharmacokinetic, and quantitative assays.

  • Deuterium-labeled internal standard for quantitative assays.
  • High purity (98.8%).
  • Soluble in DMSO at ~5 mg/mL; requires ultrasonic warming.
  • Recommended storage under nitrogen; in solution -80°C for up to 6 months, -20°C for up to 1 month.
  • Supplied in a small, analytically appropriate quantity for reference use.

Catalog No. 50-002-63571 Supplier Medchemexpress LLC Supplier No. HY113493S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Pyridoxic acid-d3 is the deuterium-labeled analog of 4-pyridoxic acid, a catabolic product of vitamin B6. It is provided as a stable isotope reference standard for use in analytical, pharmacokinetic, and quantitative assays.

  • Deuterium-labeled internal standard for quantitative assays.
  • High purity (98.8%).
  • Soluble in DMSO at ~5 mg/mL; requires ultrasonic warming.
  • Recommended storage under nitrogen; in solution -80°C for up to 6 months, -20°C for up to 1 month.
  • Supplied in a small, analytically appropriate quantity for reference use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.