Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Pyridyl)acetone 98.0+%, TCI America™
SDP

Catalog No. P20391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20391G Supplier TCI America Supplier No. P20391G

missing translation for 'validMainframeMsg'

Specifications

CAS 6304-16-1
Color Yellow
Boiling Point 80°C
Molecular Formula C8H9NO
Quantity 1 g
MDL Number MFCD00129043
Synonym 4-pyridyl acetone, 1-4-pyridinyl-2-propanone, 1-pyridin-4-yl propan-2-one, 1-4-pyridyl acetone, 4-acetonylpyridine, 4-pyridinylacetone, 4-pyridylacetone, unii-r6onm9f2au, 2-propanone, 1-4-pyridinyl
InChI Key ILRVKOYYFFNXDB-UHFFFAOYSA-N
SMILES CC(=O)CC1=CC=NC=C1
IUPAC Name 1-pyridin-4-ylpropan-2-one
Molecular Weight (g/mol) 135.166
PubChem CID 80552
Formula Weight 135.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (4-Pyridyl)acetone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.