Learn More
Medchemexpress LLC 4-pyrimidine methanamine | 45588-79-2 | MFCD06796202 | 99.3% | 109.13 g/mol | C5H7N3 | 5 G
SDP
Catalog No. 5000301466
Encompass_Preferred
4-Pyrimidine methanamine (pyrimidin-4-ylmethanamine) is a small-molecule synthetic intermediate used in medicinal and pharmaceutical chemistry. It serves as a building block for the synthesis of more complex heterocyclic compounds and is supplied for research use in laboratory-scale quantities.
  • Small pyrimidine-derived primary amine
  • Commonly used as a synthetic intermediate in medicinal chemistry
  • Molecular weight 109.13 g/mol; formula C5H7N3
  • Reported purity 99.28% (typically shown as 99.3%)
  • Physical state: liquid at ambient conditions
  • Available in multiple laboratory-scale packaging options (100 mg-50 g)
Catalog No. 50-003-01466 Supplier Medchemexpress LLC Supplier No. HYW0177695G
4-Pyrimidine methanamine (pyrimidin-4-ylmethanamine) is a small-molecule synthetic intermediate used in medicinal and pharmaceutical chemistry. It serves as a building block for the synthesis of more complex heterocyclic compounds and is supplied for research use in laboratory-scale quantities.
  • Small pyrimidine-derived primary amine
  • Commonly used as a synthetic intermediate in medicinal chemistry
  • Molecular weight 109.13 g/mol; formula C5H7N3
  • Reported purity 99.28% (typically shown as 99.3%)
  • Physical state: liquid at ambient conditions
  • Available in multiple laboratory-scale packaging options (100 mg-50 g)