Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-pyrimidinecarboxylic acid, methyl ester | 2450-08-0 | MFCD00234184 | 99.0% | 138.12 | C6H6N2O2 | 500 MG
SDP

Catalog No. 5000210212
Encompass_Preferred

Methyl pyrimidine-4-carboxylate (4-pyrimidinecarboxylic acid, methyl ester) is a synthetic intermediate used in pharmaceutical and organic synthesis. It is supplied as a high-purity powder for research use.

  • High HPLC purity: 98.96% (batch COA).
  • Molecular formula C6H6N2O2, molecular weight 138.12 g/mol.
  • Typical amount: 500 mg per package.
  • Storage stability: powder - 3 years at -20°C, 2 years at 4°C; in solvent - 6 months at -80°C, 1 month at -20°C.
  • Intended for research use as a synthetic building block in medicinal chemistry and organic synthesis.

Catalog No. 50-002-10212 Supplier Medchemexpress LLC Supplier No. HYW043311500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl pyrimidine-4-carboxylate (4-pyrimidinecarboxylic acid, methyl ester) is a synthetic intermediate used in pharmaceutical and organic synthesis. It is supplied as a high-purity powder for research use.

  • High HPLC purity: 98.96% (batch COA).
  • Molecular formula C6H6N2O2, molecular weight 138.12 g/mol.
  • Typical amount: 500 mg per package.
  • Storage stability: powder - 3 years at -20°C, 2 years at 4°C; in solvent - 6 months at -80°C, 1 month at -20°C.
  • Intended for research use as a synthetic building block in medicinal chemistry and organic synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.