Learn More
Medchemexpress LLC 4-pyrimidinemethanol | 33581-98-5 | MFCD03789625 | ≥97.0% | 110.11 | C5H6N2O | 500 MG
SDP
Catalog No. 5000301400
Encompass_Preferred
Pyrimidin-4-yl-methanol (4-pyrimidinemethanol) is a small-molecule synthetic intermediate used in medicinal and pharmaceutical chemistry for constructing pyrimidine-containing compounds. It is supplied as a solid (light yellow to yellow), with molecular formula C5H6N2O, molecular weight 110.11, CAS 33581-98-5, and typical purity ≥97%. Available in multiple pack sizes for research and synthesis.
  • Useful intermediate for synthesis of pyrimidine derivatives.
  • High reported purity suitable for research and development.
  • Well-defined molecular formula and molecular weight for formulation and analysis.
  • Supplied as a solid for straightforward handling and storage.
  • Available in multiple pack sizes to support different scales of work.
Catalog No. 50-003-01400 Supplier Medchemexpress LLC Supplier No. HYW017825500MG
Pyrimidin-4-yl-methanol (4-pyrimidinemethanol) is a small-molecule synthetic intermediate used in medicinal and pharmaceutical chemistry for constructing pyrimidine-containing compounds. It is supplied as a solid (light yellow to yellow), with molecular formula C5H6N2O, molecular weight 110.11, CAS 33581-98-5, and typical purity ≥97%. Available in multiple pack sizes for research and synthesis.
  • Useful intermediate for synthesis of pyrimidine derivatives.
  • High reported purity suitable for research and development.
  • Well-defined molecular formula and molecular weight for formulation and analysis.
  • Supplied as a solid for straightforward handling and storage.
  • Available in multiple pack sizes to support different scales of work.