Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-pyrimidinemethanol | 33581-98-5 | MFCD03789625 | >97.0% | 110.11 g/mol | C5H6N2O | 1 G
SDP

Catalog No. 5000301310
Encompass_Preferred

Pyrimidin-4-yl-methanol is a small-molecule synthetic intermediate used in medicinal chemistry and agrochemical research. It is provided as a solid, high-purity reagent for use as a building block in heterocyclic synthesis and other organic transformations.

  • High purity (≥97.0%).
  • Molecular weight 110.11 g/mol.
  • Chemical formula: C5H6N2O.
  • Solid, light yellow to yellow physical form.
  • Supplied in laboratory-scale package sizes (typically 1 g).
  • Suitable as a synthetic intermediate for pharmaceutical and agrochemical synthesis.

Catalog No. 50-003-01310 Supplier Medchemexpress LLC Supplier No. HYW0178251G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrimidin-4-yl-methanol is a small-molecule synthetic intermediate used in medicinal chemistry and agrochemical research. It is provided as a solid, high-purity reagent for use as a building block in heterocyclic synthesis and other organic transformations.

  • High purity (≥97.0%).
  • Molecular weight 110.11 g/mol.
  • Chemical formula: C5H6N2O.
  • Solid, light yellow to yellow physical form.
  • Supplied in laboratory-scale package sizes (typically 1 g).
  • Suitable as a synthetic intermediate for pharmaceutical and agrochemical synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.