Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride

Catalog No. CDS014683 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014683 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014683 Supplier Sigma Aldrich Supplier No. CDS014683100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-methoxy-1-propanol is a primary alcohol
  • Kinetics of the reactions of 3-methoxy-1-propanol with phenyl isocyanate in the presence of metal catalysts has been reported
  • Selective synthesis of 3-methoxy-1-propanol from methanol and allyl alcohol with metal oxides and zeolites catalysts in the liquid phase has been studied

Specifications

Molecular Formula C10H12ClNO4S2
Linear Formula C10H12ClNO4S2
MDL Number MFCD03407360
Quantity 10 mL
Molecular Weight (g/mol) 309.79
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.