Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-quinolinecarboxylic acid, 7-bromo- | 31009-04-8 | MFCD11108751 | 96.1% | 252.06 | C10H6BrNO2 | 250 MG
SDP

Catalog No. 5000302378
Encompass_Preferred

7-Bromoquinoline-4-carboxylic acid is an aromatic heterocyclic carboxylic acid used as an intermediate and building block in organic synthesis, particularly in medicinal chemistry and materials research. It is typically handled as a solid reagent and supplied with batch documentation for quality control.

  • Used as an intermediate in organic and medicinal chemistry.
  • HPLC purity 96.1%.
  • Molecular formula C10H6BrNO2; molecular weight 252.06.
  • Appearance: gray to brown solid.
  • Storage: powder -20°C (stable 3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Batch certificate of analysis and safety data sheet available.

Catalog No. 50-003-02378 Supplier Medchemexpress LLC Supplier No. 250MGHYW022164
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Bromoquinoline-4-carboxylic acid is an aromatic heterocyclic carboxylic acid used as an intermediate and building block in organic synthesis, particularly in medicinal chemistry and materials research. It is typically handled as a solid reagent and supplied with batch documentation for quality control.

  • Used as an intermediate in organic and medicinal chemistry.
  • HPLC purity 96.1%.
  • Molecular formula C10H6BrNO2; molecular weight 252.06.
  • Appearance: gray to brown solid.
  • Storage: powder -20°C (stable 3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Batch certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.