Learn More
Apexbio Technology LLC 4-quinolinol 611-36-9 100mg
SDP
Catalog No. 5000090957
Encompass_Preferred
4-Quinolinol (CAS 611-36-9) is a hydroxylated derivative of quinoline with demonstrated antimicrobial properties It exerts inhibitory effects on the growth of various intestinal bacteria suggesting interference with microbial viability or metabolic pathways Additionally 4-quinolinol has been utilized in research as a sacrificial scavenger for photogenerated reactive oxygen species supporting studies on oxidative stress and photochemical processes These features highlight its utility in microbiological research and investigations into oxidative mechanisms
Catalog No. 50-000-90957 Supplier Apexbio Technology LLC Supplier No. B7871100
4-Quinolinol (CAS 611-36-9) is a hydroxylated derivative of quinoline with demonstrated antimicrobial properties It exerts inhibitory effects on the growth of various intestinal bacteria suggesting interference with microbial viability or metabolic pathways Additionally 4-quinolinol has been utilized in research as a sacrificial scavenger for photogenerated reactive oxygen species supporting studies on oxidative stress and photochemical processes These features highlight its utility in microbiological research and investigations into oxidative mechanisms