Learn More
Medchemexpress LLC 4-Quinolinol | 611-36-9 | 99.9% | C9H7NO | 1 ML
SDP
Catalog No. 5000434001
Encompass_Preferred
4-Quinolinol (Kynurine) is a quinoline derivative that modulates tryptophan metabolism through the Kynurine pathway, involved in neuroprotective effects. It promotes tumor cell survival and motility by suppressing antitumor immune responses, particularly active in human brain tumors where AhR activation enhances TDO expression, producing more kynurine and kynurinic acid.
  • Quinoline derivative
  • Modulates tryptophan metabolism
  • Involved in neuroprotective effects
  • Promotes tumor cell survival and motility
  • Suppresses antitumor immune responses
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-004-34001 Supplier Medchemexpress LLC Supplier No. HY5920810MM/1ML
4-Quinolinol (Kynurine) is a quinoline derivative that modulates tryptophan metabolism through the Kynurine pathway, involved in neuroprotective effects. It promotes tumor cell survival and motility by suppressing antitumor immune responses, particularly active in human brain tumors where AhR activation enhances TDO expression, producing more kynurine and kynurinic acid.
  • Quinoline derivative
  • Modulates tryptophan metabolism
  • Involved in neuroprotective effects
  • Promotes tumor cell survival and motility
  • Suppresses antitumor immune responses
  • Soluble in DMSO (100 mg/mL)