Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-((R)-1-aminoethyl)-N-(pyridin-4-yl)benzamide | 199433-58-4 | 98.4% | 280.33 | C16H16N4O | 1 ML
SDP

Catalog No. 5000214188
Encompass_Preferred

Y-33075 is a selective Rho-associated kinase (ROCK) inhibitor derived from Y-27632. It is reported to be more potent than Y-27632, with an IC50 of approximately 3.6 nM, and is used in biochemical and cellular studies to investigate ROCK-dependent signaling.

  • Selective ROCK inhibition with low-nanomolar potency (IC50 ≈ 3.6 nM).
  • Useful for biochemical and cellular assays probing ROCK signaling.
  • High chemical purity (~98.4%) suitable for research use.
  • Known chemical identity: 4-((R)-1-aminoethyl)-N-(pyridin-4-yl)benzamide, C16H16N4O.
  • Molecular weight 280.33 g/mol.

Catalog No. 50-002-14188 Supplier Medchemexpress LLC Supplier No. HY1006710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Y-33075 is a selective Rho-associated kinase (ROCK) inhibitor derived from Y-27632. It is reported to be more potent than Y-27632, with an IC50 of approximately 3.6 nM, and is used in biochemical and cellular studies to investigate ROCK-dependent signaling.

  • Selective ROCK inhibition with low-nanomolar potency (IC50 ≈ 3.6 nM).
  • Useful for biochemical and cellular assays probing ROCK signaling.
  • High chemical purity (~98.4%) suitable for research use.
  • Known chemical identity: 4-((R)-1-aminoethyl)-N-(pyridin-4-yl)benzamide, C16H16N4O.
  • Molecular weight 280.33 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.