Learn More
Chemglass Life Sciences Reaction Kettle Flange, 102mm(4in) ID
SDP
Catalog No. CG14301
Encompass_Preferred
Reaction Kettle Flange, 102mm(4in) ID. Conical style flange has an I.D. of 4in with a flat, ground surface used for construction of reaction kettles or flasks. Tooled on tubing having an I.D. of 105mm and an O.D. of 115mm. Supplied with approximately 7in of tubing below joint. Available tooled on longer lengths of tubing. Please contact us for details.
Catalog No. CG14301 Supplier Chemglass Life Sciences Supplier No. CG14301
Reaction Kettle Flange, 102mm(4in) ID. Conical style flange has an I.D. of 4in with a flat, ground surface used for construction of reaction kettles or flasks. Tooled on tubing having an I.D. of 105mm and an O.D. of 115mm. Supplied with approximately 7in of tubing below joint. Available tooled on longer lengths of tubing. Please contact us for details.