Learn More
Chemglass Life Sciences 4 Ring Hose Connection, "A" = 5.0, "B" = 12.0, "C" = 14, "D" = 24; "E" = 12
SDP
Catalog No. 501531985
Encompass_Preferred
4 Ring Hose Connection, "A" = 5.0, "B" = 12.0, "C" = 14, "D" = 24; "E" = 12. Connections are tooled to form a flare for sealing directly to apparatus using the tubing as a holder. Serrations are tooled on both ends of an 8" length of tubing.
Catalog No. 50-153-1985 Supplier Chemglass Life Sciences Supplier No. CGB30102
4 Ring Hose Connection, "A" = 5.0, "B" = 12.0, "C" = 14, "D" = 24; "E" = 12. Connections are tooled to form a flare for sealing directly to apparatus using the tubing as a holder. Serrations are tooled on both ends of an 8" length of tubing.