Learn More
Chemglass Life Sciences 4 Ring Hose Connection, "A" = 8.5, "B" = 13.5, "C" = 15.5, "D" = 20; "E" = 12
SDP
Catalog No. CG30003
Encompass_Preferred
4 Ring Hose Connection, "A" = 8.5, "B" = 13.5, "C" = 15.5, "D" = 20; "E" = 12. Serrations are tooled on both ends of an 8" length of tubing. Hose connections are SOLD IN PACKAGES OF 10 SERRATED CONNECTORS (5 pieces tooled on both ends).
Catalog No. CG30003 Supplier Chemglass Life Sciences Supplier No. CG30003
4 Ring Hose Connection, "A" = 8.5, "B" = 13.5, "C" = 15.5, "D" = 20; "E" = 12. Serrations are tooled on both ends of an 8" length of tubing. Hose connections are SOLD IN PACKAGES OF 10 SERRATED CONNECTORS (5 pieces tooled on both ends).