Learn More
New Era Pump Systems Inc 4 roller 1/16in ID tubing kit green 4 roller head - 4 ft. of PharMed tubing - 8in nylon clamp ties reduction couplers and a vial of lubricant
Catalog No. NC3343305
Encompass
4 roller 1/16in ID tubing kit green 4 roller head - 4 ft. of PharMed tubing - 8in nylon clamp ties reduction couplers and a vial of lubricant
Catalog No. NC3343305 Supplier New Era Pump Systems Inc Supplier No. PERIHEADKITYG4
4 roller 1/16in ID tubing kit green 4 roller head - 4 ft. of PharMed tubing - 8in nylon clamp ties reduction couplers and a vial of lubricant