Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-((S)-2-((S)-2-(6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamido)-3-methylbutanamido)-5-ur... | 2722697-82-5 | MFCD00867744 | 99.9% | 1126.65 g·mol-1 | C55H68ClN11O13 | 5 MG
SDP

Catalog No. 5000221658
Encompass_Preferred

MC-VC-PABC-amide-PEG1-CH2-CC-885 is a neoDegrader-linker conjugate that couples a GSPT1-targeting degrader payload to a cleavable linker via a short PEG spacer. It is supplied as a research reagent for biochemical and cell-based studies of targeted protein degradation and assay development.

  • NeoDegrader-linker conjugate designed for targeted protein degradation studies.
  • GSPT1-targeting degrader payload for selective biological activity.
  • Cleavable valine-citrulline PABC-type linker enabling intracellular payload release.
  • Short PEG spacer improves solubility and linker flexibility.
  • High reported purity suitable for research applications.
  • Soluble in DMSO; may require sonication, use anhydrous DMSO for best results.
  • Store at -20°C, protect from light; in solvent store at -80°C for long-term storage.

Catalog No. 50-002-21658 Supplier Medchemexpress LLC Supplier No. HY1454485MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MC-VC-PABC-amide-PEG1-CH2-CC-885 is a neoDegrader-linker conjugate that couples a GSPT1-targeting degrader payload to a cleavable linker via a short PEG spacer. It is supplied as a research reagent for biochemical and cell-based studies of targeted protein degradation and assay development.

  • NeoDegrader-linker conjugate designed for targeted protein degradation studies.
  • GSPT1-targeting degrader payload for selective biological activity.
  • Cleavable valine-citrulline PABC-type linker enabling intracellular payload release.
  • Short PEG spacer improves solubility and linker flexibility.
  • High reported purity suitable for research applications.
  • Soluble in DMSO; may require sonication, use anhydrous DMSO for best results.
  • Store at -20°C, protect from light; in solvent store at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.