Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-((S)-4-acryloyl-2-methylpiperazin-1-yl)-6-fluoro-7-(2-fluoro-6-hydroxyphenyl)-1-(2-isopropyl-4-methy | 00-00-0 | 99.1% | 560.59 g/mol | C30H30F2N6O3 | 5 MG
SDP

Catalog No. 5000264509
Encompass_Preferred

Sotorasib isomer is the less-active stereoisomer of the KRAS G12C covalent inhibitor sotorasib. Supplied for research use only, it is intended for in vitro and in vivo studies of Ras signaling and KRAS G12C biology, with analytical and storage data provided to guide experimental handling.

  • Purity: 99.1% (LCMS).
  • Molecular weight: 560.59 g/mol.
  • Chemical formula: C30H30F2N6O3.
  • Storage: powder at -20°C (up to 3 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Solubility: H2O <0.1 mg/mL (insoluble); ≥1.25 mg/mL in 10% DMSO for in vivo use.
  • Target: Ras (KRAS G12C) pathway support for MAPK/ERK studies.
  • Intended use: for research use only; not for human or clinical use.

Catalog No. 50-002-64509 Supplier Medchemexpress LLC Supplier No. HY114277B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sotorasib isomer is the less-active stereoisomer of the KRAS G12C covalent inhibitor sotorasib. Supplied for research use only, it is intended for in vitro and in vivo studies of Ras signaling and KRAS G12C biology, with analytical and storage data provided to guide experimental handling.

  • Purity: 99.1% (LCMS).
  • Molecular weight: 560.59 g/mol.
  • Chemical formula: C30H30F2N6O3.
  • Storage: powder at -20°C (up to 3 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Solubility: H2O <0.1 mg/mL (insoluble); ≥1.25 mg/mL in 10% DMSO for in vivo use.
  • Target: Ras (KRAS G12C) pathway support for MAPK/ERK studies.
  • Intended use: for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.