Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[(S)-amino(carboxy)methyl]-3-methylbenzoic acid hydrochloride | 2829282-00-8 | MFCD02262124 | 99.4% | 245.66 g/mol | C10H12ClNO4 | 1 ML
SDP

Catalog No. 5000241449
Encompass_Preferred

LY367385 hydrochloride is a selective mGluR1a antagonist supplied as a hydrochloride salt for research use. It is available as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL vial), suitable for in vitro pharmacology and biochemical assays.

  • High purity (99.4%) for reproducible assay results.
  • Available as 10 mM solution in DMSO for convenient dosing.
  • Also supplied as solid for custom formulation and storage.
  • Molecular weight 245.66 g/mol and formula C10H12ClNO4.
  • CAS number 2829282-00-8 for unambiguous substance identification.

Catalog No. 50-002-41449 Supplier Medchemexpress LLC Supplier No. HY107515A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY367385 hydrochloride is a selective mGluR1a antagonist supplied as a hydrochloride salt for research use. It is available as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL vial), suitable for in vitro pharmacology and biochemical assays.

  • High purity (99.4%) for reproducible assay results.
  • Available as 10 mM solution in DMSO for convenient dosing.
  • Also supplied as solid for custom formulation and storage.
  • Molecular weight 245.66 g/mol and formula C10H12ClNO4.
  • CAS number 2829282-00-8 for unambiguous substance identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.