Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex 4 Slot Cell Culture Plate Organizer
SDP

Catalog No. 22000440
Change view
Click to view available options
Positions:
4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
22-000-440 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-000-440 Supplier Poltex Supplier No. CELLCUL4
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Organizes cell culture plates.

Durable and cleanable PETG construction

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Cell Culture, Elisa Plates
Material PETG
Positions 4
Sterility Non-sterile
For Use With (Equipment) Cell culture tray, petri dish
Quantity 1
Length (Metric) 185 mm
Weight (Metric) 1360 g
Includes Rack
Height (Metric) 360 mm
Width (Metric) 489 mm
Show More Show Less

Consumables not included.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.