Learn More
Bellco Glass, Inc. 4" Soft Rubber Plate Roller
SDP
Catalog No. 501945612
Encompass_Preferred
4" Soft Rubber Plate Roller . Ideal use for sealing films to microplates.
Catalog No. 50-194-5612 Supplier Bellco Glass, Inc. Supplier No. 440001500
4" Soft Rubber Plate Roller . Ideal use for sealing films to microplates.